MAN375328153 Manchester Organics

2-Bromo-4-chloro-1-(trifluoromethoxy)benzene

  • MDL number MFCD11847191
  • Molecular Weight : 275.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/X15960
Availability details for
Quantity
  • Availability
  • Add to Cart