MAN375328432 Manchester Organics

5-Bromo-1,3-dichloro-2-(trifluoromethoxy)benzene

  • MDL number MFCD22571071
  • Molecular Weight : 309.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F14218
Availability details for
Quantity
  • Availability
  • Add to Cart