MAN375328720 Manchester Organics

Sodium difluoroacetate

  • MDL number MFCD00066171
  • Molecular Weight : 118.01471

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A27774

Safety Information

MSDS https://www.manchesterorganics.com/A27774
Availability details for
Quantity
  • Availability
  • Add to Cart