MAN375328749 Manchester Organics

1-(2,3,4,5-Tetrafluorophenyl)ethanone

  • MDL number MFCD00134310
  • Cas Number : 66286-21-3
  • Molecular Weight : 192.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/T27790
Availability details for
Quantity
  • Availability
  • Add to Cart