MAN375329019 Manchester Organics

2-(4-Methylpiperazin-1-yl)acetic acid

  • MDL number MFCD03444554
  • Molecular Weight : 158.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/N33731
Availability details for
Quantity
  • Availability
  • Add to Cart