MAN375329067 Manchester Organics

3-(Difluoromethyl)-1-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD11865267
  • Molecular Weight : 176.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C27847
Availability details for
Quantity
  • Availability
  • Add to Cart