MAN375329118 Manchester Organics

2-Fluoro-1,3,5-trimethyl-benzene

  • MDL number MFCD00052365
  • Cas Number : 392-69-8
  • Molecular Weight : 138.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin GB
Product Link https://www.manchesterorganics.com/A33811
Availability details for
Quantity
  • Availability
  • Add to Cart