MAN375329243 Manchester Organics

Ethyl 2-amino-4-methyl-thiophene-3-carboxylate

  • MDL number MFCD00051669
  • Cas Number : 43088-42-2
  • Molecular Weight : 185.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin GB
Product Link https://www.manchesterorganics.com/E33907
Availability details for
Quantity
  • Availability
  • Add to Cart