MAN375329330 Manchester Organics

(1,5-Dimethylpyrazol-3-yl)methanamine

  • MDL number MFCD03422531
  • Cas Number : 423768-52-9
  • Molecular Weight : 125.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/A33995
Availability details for
Quantity
  • Availability
  • Add to Cart