MAN375329610 Manchester Organics

[3-(Trifluoromethyl)phenyl] carbonochloridate

  • MDL number MFCD02093719
  • Cas Number : 95668-29-4
  • Molecular Weight : 224.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin GB
Product Link https://www.manchesterorganics.com/G34047
Availability details for
Quantity
  • Availability
  • Add to Cart