MAN375329696 Manchester Organics

3-[5-(2-Fluorophenyl)-1,2,4-oxadiazol-3-yl]benzoic acid

  • MDL number MFCD09864996
  • Cas Number : 775304-57-9
  • Molecular Weight : 284.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/A34202
Availability details for
Quantity
  • Availability
  • Add to Cart