MAN412111902 Manchester Organics

Cyclohexyl-(4-methoxyphenyl)methanone

  • MDL number MFCD03841341
  • Molecular Weight : 218.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y34315
Availability details for
Quantity
  • Availability
  • Add to Cart