MAN412111996 Manchester Organics

5-(Aminomethyl)-1,3-dimethyl-1,3-dihydro-2H-benzimidazol-2-one hydrochloride

  • MDL number MFCD05022455
  • Molecular Weight : 227.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F20347

Safety Information

MSDS https://www.manchesterorganics.com/F20347
Availability details for
Quantity
  • Availability
  • Add to Cart