MAN412111998 Manchester Organics

2-Bromo-5-methoxyaniline hydrochloride

  • MDL number MFCD10699672
  • Molecular Weight : 238.50

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A20626

Safety Information

MSDS https://www.manchesterorganics.com/A20626
Availability details for
Quantity
  • Availability
  • Add to Cart