MAN412112049 Manchester Organics

3-Bromo-2-chloro-5-nitro-6-picoline

  • MDL number MFCD09839270
  • Cas Number : 856834-95-2
  • Molecular Weight : 251.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/W28597
Availability details for
Quantity
  • Availability
  • Add to Cart