MAN412112055 Manchester Organics

Methyl 4-hydroxypyrrolidine-2-carboxylate

  • MDL number MFCD04614645
  • Molecular Weight : 145.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin GB
Product Link https://www.manchesterorganics.com/J28137
Availability details for
Quantity
  • Availability
  • Add to Cart