MAN412112101 Manchester Organics

4-Aminocyclohexanecarboxylic acid hydrochloride

  • MDL number MFCD00800477
  • Cas Number : 854446-76-7
  • Molecular Weight : 179.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-aminocyclohexanecarboxylic acid hydrochloride
Inchi Code 1S/C7H13NO2.ClH/c8-6-3-1-5(2-4-6)7(9)10;/h5-6H,1-4,8H2,(H,9,10);1H
Inchi Key HXZSYUOXTKQNNY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/E12087
Availability details for
Quantity
  • Availability
  • Add to Cart