MAN412112183 Manchester Organics

1,3,5-Trifluoro-2,4,6-triiodobenzene

  • MDL number MFCD06248899
  • Cas Number : 84322-56-5
  • Molecular Weight : 509.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/K28232
Availability details for
Quantity
  • Availability
  • Add to Cart