MAN412112289 Manchester Organics

4,4-Difluoropiperidine-1-sulfonyl chloride

  • MDL number MFCD18917106
  • Cas Number : 1224929-81-0
  • Molecular Weight : 219.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4,4-difluoro-1-piperidinesulfonyl chloride
Inchi Code 1S/C5H8ClF2NO2S/c6-12(10,11)9-3-1-5(7,8)2-4-9/h1-4H2
Inchi Key QSGQFPJNSNFIMU-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/D14690
Availability details for
Quantity
  • Availability
  • Add to Cart