MAN412112297 Manchester Organics

4,4,4-Trifluoro-3-hydroxy-1-phenylbutan-1-one

  • MDL number MFCD07779795
  • Molecular Weight : 218.172

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 0.95
Availability details for
Quantity
  • Availability
  • Add to Cart