MAN412112363 Manchester Organics

Trifluoromethylsulfanyl-acetic acid ethyl ester

  • MDL number MFCD16617909
  • Molecular Weight : 188.168

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 0.97
Availability details for
Quantity
  • Availability
  • Add to Cart