MAN412112552 Manchester Organics

2,2'-Biimidazole

  • MDL number MFCD00047014
  • Molecular Weight : 134.13864

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A12248

Safety Information

MSDS https://www.manchesterorganics.com/A12248
Availability details for
Quantity
  • Availability
  • Add to Cart