MAN412112644 Manchester Organics

3,4,5-Trifluoropyridine

  • MDL number MFCD09998883
  • Molecular Weight : 133.071

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/U28714
Availability details for
Quantity
  • Availability
  • Add to Cart