MAN412112812 Manchester Organics

4-Bromo-2-(difluoromethyl)pyridine hydrobromide

  • MDL number MFCD23382408
  • Cas Number : 1422766-41-3
  • Molecular Weight : 288.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/F12253
Availability details for
Quantity
  • Availability
  • Add to Cart