MAN412113061 Manchester Organics

Benzyl 3-fluoro-4-oxo-piperidine-1-carboxylate

  • MDL number MFCD11976639
  • Cas Number : 845256-59-9
  • Molecular Weight : 251.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/L12258
Availability details for
Quantity
  • Availability
  • Add to Cart