MAN412113153 Manchester Organics

Tetradecanal

  • MDL number MFCD00007019
  • Molecular Weight : 212.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C34572
Availability details for
Quantity
  • Availability
  • Add to Cart