MAN412113237 Manchester Organics

4-(3-Methyloxetan-3-yl)aniline

  • MDL number MFCD20486517
  • Cas Number : 1225380-12-0
  • Molecular Weight : 163.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(3-methyl-3-oxetanyl)aniline
Inchi Code 1S/C10H13NO/c1-10(6-12-7-10)8-2-4-9(11)5-3-8/h2-5H,6-7,11H2,1H3
Inchi Key VLCQTNSZCIBOTL-UHFFFAOYSA-N

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/C34595
Availability details for
Quantity
  • Availability
  • Add to Cart