MAN412113259 Manchester Organics

2-Bromo-5-methylfuran

  • MDL number MFCD11870159
  • Molecular Weight : 160.997

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/T34609
Availability details for
Quantity
  • Availability
  • Add to Cart