MAN412113295 Manchester Organics

Methyl 6-chloro-4-(trifluoromethyl)nicotinate

  • MDL number MFCD01941315
  • Molecular Weight : 239.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B14498
Availability details for
Quantity
  • Availability
  • Add to Cart