MAN412113309 Manchester Organics

4-Bromo-3-chlorophenol

  • MDL number MFCD00070744
  • Molecular Weight : 207.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/W34635
Availability details for
Quantity
  • Availability
  • Add to Cart