MAN412113354 Manchester Organics

2-[3,6-Bis(dimethylamino)xanthen-10-ium-9-yl]-5-carboxy-benzoate

  • MDL number MFCD00269769
  • Molecular Weight : 430.453

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/K34671
Availability details for
Quantity
  • Availability
  • Add to Cart