MAN412113367 Manchester Organics

4,5-Dihydroxynaphthalene-2,7-disulfonic acid

  • MDL number MFCD00003968
  • Cas Number : 148-25-4
  • Molecular Weight : 320.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 75
Country of Origin GB
Product Link https://www.manchesterorganics.com/B34686
Availability details for
Quantity
  • Availability
  • Add to Cart