MAN412113387 Manchester Organics

4-Bromo-5-methoxy-1H-indole

  • MDL number MFCD12973810
  • Cas Number : 90858-86-9
  • Molecular Weight : 226.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-5-methoxy-1H-indole
Inchi Code 1S/C9H8BrNO/c1-12-8-3-2-7-6(9(8)10)4-5-11-7/h2-5,11H,1H3
Inchi Key FPSVUEYKYLKRJL-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/N34697
Availability details for
Quantity
  • Availability
  • Add to Cart