MAN412113452 Manchester Organics

4-Chloro-2-(trifluoromethyl)pyrimidine

  • MDL number MFCD06411632
  • Molecular Weight : 182.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin GB
Product Link https://www.manchesterorganics.com/T12264
Availability details for
Quantity
  • Availability
  • Add to Cart