MAN412113461 Manchester Organics

1-Benzhydryl-3-iodo-azetidine

  • MDL number MFCD08062427
  • Cas Number : 125735-40-2
  • Molecular Weight : 349.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin GB
Product Link https://www.manchesterorganics.com/F34759
Availability details for
Quantity
  • Availability
  • Add to Cart