MAN412113492 Manchester Organics

4-Bromo-2,6-dichlorophenol

  • MDL number MFCD00019996
  • Cas Number : 3217-15-0
  • Molecular Weight : 241.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-2,6-dichlorophenol
Inchi Code 1S/C6H3BrCl2O/c7-3-1-4(8)6(10)5(9)2-3/h1-2,10H
Inchi Key KGURSDWHGSLAPP-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/M34788
Availability details for
Quantity
  • Availability
  • Add to Cart