MAN412113524 Manchester Organics

3-Bromo-4-nitropyridine

  • MDL number MFCD03085767
  • Cas Number : 89364-04-5
  • Molecular Weight : 203

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-4-nitropyridine
Inchi Code 1S/C5H3BrN2O2/c6-4-3-7-2-1-5(4)8(9)10/h1-3H
Inchi Key CJEWCWSWUUFORD-UHFFFAOYSA-N

Description

Purity 96%
Country of Origin GB
Product Link https://www.manchesterorganics.com/V34818
Availability details for
Quantity
  • Availability
  • Add to Cart