MAN412113526 Manchester Organics

1,3-Benzothiazole-7-carboxylic acid

  • MDL number MFCD13195419
  • Cas Number : 677304-83-5
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/X34820
Availability details for
Quantity
  • Availability
  • Add to Cart