MAN412113538 Manchester Organics

2-Chloro-1,3-benzothiazole-6-carbonitrile

  • MDL number MFCD09743933
  • Cas Number : 80945-83-1
  • Molecular Weight : 194.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-1,3-benzothiazole-6-carbonitrile
Inchi Code 1S/C8H3ClN2S/c9-8-11-6-2-1-5(4-10)3-7(6)12-8/h1-3H
Inchi Key DHBTVASVKVLZBJ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/K34832
Availability details for
Quantity
  • Availability
  • Add to Cart