MAN412113609 Manchester Organics

4-(1-Phenylethyl)benzene-1,3-diol

  • MDL number MFCD21648466
  • Cas Number : 85-27-8
  • Molecular Weight : 214.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin GB
Product Link https://www.manchesterorganics.com/S34907
Availability details for
Quantity
  • Availability
  • Add to Cart