MAN412113643 Manchester Organics

4-(Trifluoromethyl)-1H-indole

  • MDL number MFCD09954766
  • Cas Number : 128562-95-8
  • Molecular Weight : 185.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/G34944
Availability details for
Quantity
  • Availability
  • Add to Cart