MAN412113683 Manchester Organics

Methyl 5-fluoropyridine-2-carboxylate

  • MDL number MFCD11845630
  • Molecular Weight : 155.12644

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B34985

Safety Information

MSDS https://www.manchesterorganics.com/B34985
Availability details for
Quantity
  • Availability
  • Add to Cart