MAN412113762 Manchester Organics

Ethyl 4-amino-6-chloro-pyridine-3-carboxylate

  • MDL number MFCD00834975
  • Cas Number : 380626-81-3
  • Molecular Weight : 200.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/S35068
Availability details for
Quantity
  • Availability
  • Add to Cart