MAN412113772 Manchester Organics

2-Hydroxy-5-methyl-pyridine-3-carboxylic acid

  • MDL number MFCD11227183
  • Cas Number : 38076-77-6
  • Molecular Weight : 153.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/E11070
Availability details for
Quantity
  • Availability
  • Add to Cart