MAN412113810 Manchester Organics

8-Bromooct-1-yn-3-ol

  • MDL number MFCD24465304
  • Cas Number : 1579278-86-6
  • Molecular Weight : 205.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/D11069
Availability details for
Quantity
  • Availability
  • Add to Cart