MAN412113830 Manchester Organics

Prop-2-ene-1-sulfonyl chloride

  • MDL number MFCD08059789
  • Molecular Weight : 140.5886

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/M35133

Safety Information

MSDS https://www.manchesterorganics.com/M35133
Availability details for
Quantity
  • Availability
  • Add to Cart