MAN412113970 Manchester Organics

Ethyl 5-methyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD00233455
  • Cas Number : 4027-57-0
  • Molecular Weight : 154.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 5-methyl-1H-pyrazole-3-carboxylate
Inchi Code 1S/C7H10N2O2/c1-3-11-7(10)6-4-5(2)8-9-6/h4H,3H2,1-2H3,(H,8,9)
Inchi Key BOTXQJAHRCGJEG-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/D35332
Availability details for
Quantity
  • Availability
  • Add to Cart