MAN412114219 Manchester Organics

(2S)-N-(2,6-Dimethylphenyl)-1-propyl-piperidine-2-carboxamide

  • MDL number MFCD00864425
  • Molecular Weight : 274.401

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A35582
Availability details for
Quantity
  • Availability
  • Add to Cart