MAN412114426 Manchester Organics

1-Methyl-1H-pyrazol-4-ylamine xHCl

  • MDL number MFCD07626616
  • Cas Number : 127107-23-7
  • Molecular Weight : 170.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-methyl-1H-pyrazol-4-amine dihydrochloride
Inchi Code 1S/C4H7N3.2ClH/c1-7-3-4(5)2-6-7;;/h2-3H,5H2,1H3;2*1H
Inchi Key HEFHQCFTJGKPMD-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/Y21854
Availability details for
Quantity
  • Availability
  • Add to Cart