MAN412114481 Manchester Organics

1,6,7,12-Tetrachloroperylene tetracarboxylic acid dianhydride

  • MDL number MFCD11870880
  • Cas Number : 156028-26-1
  • Molecular Weight : 530.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/D35907
Availability details for
Quantity
  • Availability
  • Add to Cart